Orders LinkContactUsLinkComponent
Lab Consumables
Lab Consumables
Plated brass loop holder with PVC handle, 175 mm, 100×1, in tube
Katalog # 612-2686
Leverandør:   Copan
Lab Consumables
Katalog # 612-2686
Leverandør:  Copan
Lab Consumables
Lab Consumables
Plated brass loop holder with PVC handle, 175 mm, 100×1, in tube
Price
333,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
A variety of high-quality lab consumables to meet laboratories everyday needs. We offer a wide range of transfer pipets, loops, spreaders, tubes, caps, and droppers meeting the highest quality standards. Since we produce them with our in-house technology, we can easily customise them upon customer’s request.
Streaking:
Loops and spreaders
Needles, spreaders, and inoculating loops fitting a multitude of different tasks
- Lubricants, oils, and electrostatic charges free
- Certified calibration
- Hexagonal loop shaft for easy handling
- Evans blue dye accuracy certification
Specifikationer
  • Pakket:
    100×1, in tube
  • Beskrivelse:
    Plated brass loop holder with PVC handle, 175 mm
  • Seq:
    4
  • Cat. No.:
    612-2686