VWR®, Microscope Slide Holders
Objektglasholder til 40 objektglas, hvid, PE, 178×299×21 mm, 40
Katalog # 631-0718
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Sturdy microscope slide holder designed for drying and cleaning.
Colour: White
Documents
VWR® Collection Selection Catalogue
Produktlitteratur
Specifikationer
- Antal objektglas:40
- Farve:hvid
- Beskrivelse:Objektglasholder til 40 objektglas
- Materiale:PE
- Seq:03
- B×D×H:178×299×21 mm