VWR®, Microscope Slide Holders
Objektglasholder til 15 objektglas, hvid, PE, 102×153×21 mm, 15
Katalog # 631-0717
Leverandør: VWR Collection
Gå til:
Product Details & Documents
Sturdy microscope slide holder designed for drying and cleaning.
Colour: White
Documents
VWR® Collection Selection Catalogue
Produktlitteratur
Specifikationer
- Antal objektglas:15
- Farve:hvid
- Beskrivelse:Objektglasholder til 15 objektglas
- Materiale:PE
- Seq:01
- B×D×H:102×153×21 mm