VWR®, Transfer Pipettes, High Performance
1,5 ml, 28, 1,0 ml, 5 mm, 123 mm, - ml, -, Bulk
Leverandør: VWR Collection
Gå til:
Product Details & Documents
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
- Non toxic and inert to biological fluids and most acids
- Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
- Available sterile or non sterile
- Sterility assurance level is SAL 10⁻⁶
Emballering: Sterilt-emballerede pipetter i papir i medicinsk kvalitet og ugiftig film.
Documents
Centrifugation Solutions from Avantor VWR® for Liquid Handling
Produktlitteratur
CoC VWR Disposable High Performance Transfer Pipettes.pdf CoC VWR Transfer Pipettes, High-Performance.pdf
Overensstemmelseserklæring
Specifikationer
- Pakket:Bulk
- Steril:-
- Kapacitet:1,5 ml
- Længde:123 mm
- Seq:02
- Dråber pr ml:28
- Inddeling:- ml
- Stilk Ø:5 mm
- Sugevolumen:1,0 ml
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



