Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VWR®, Transfer Pipettes, High Performance

VWR®, Transfer Pipettes, High Performance

14,0 ml, 18, 10,0 ml, 4 mm, 170 mm, - ml, -, Bulk
Katalog # 612-2847
Leverandør:   VWR Collection
VWR®, Transfer Pipettes, High Performance
Katalog # 612-2847
Leverandør:  VWR Collection
VWR®, Transfer Pipettes, High Performance
VWR®, Transfer Pipettes, High Performance

14,0 ml, 18, 10,0 ml, 4 mm, 170 mm, - ml, -, Bulk

Price
236,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents

LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.

  • Non toxic and inert to biological fluids and most acids
  • Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
  • Available sterile or non sterile
  • Sterility assurance level is SAL 10⁻⁶

Emballering: Sterilt-emballerede pipetter i papir i medicinsk kvalitet og ugiftig film.

Specifikationer
  • Pakket:
    Bulk
  • Steril:
    -
  • Kapacitet:
    14,0 ml
  • Længde:
    170 mm
  • Seq:
    17
  • Dråber pr ml:
    18
  • Inddeling:
    - ml
  • Stilk Ø:
    4 mm
  • Sugevolumen:
    10,0 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.