Sample cups for analysers, ratiolab®
For Hitachi H1 analysers, 3 ml
Catalog # RATI5700401
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Made of clear PS.
- Good chemical resistance
- Good temperature resistance
Analysis beakers offer the highest precision for use with robotic analytical systems.
Specifications
- Kapacitet:3 ml
- Beskrivelse:For Hitachi H1 analysers
- Seq:03