Probes for 845
TC K-type, fast action surface probe with sprung thermocouple strip for uneven surfaces, −60…+300 °C, ±0,3 (−60…+60); ±0,5 °C, 5×115 mm
Katalog # 620-1207
Leverandør: Testo
Gå til:
Product Details & Documents
Specifikationer
- Område:−60…+300 °C
- Beskrivelse:TC K-type, fast action surface probe with sprung thermocouple strip for uneven surfaces
- Seq:22
- Nøjagtighed:±0,3 (−60…+60); ±0,5 °C
- Ø×L:5×115 mm