pH probes
Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 11×35 mm, Semi-solid substances
Katalog # TEST0650.2062
Leverandør: Testo
Gå til:
Product Details & Documents
Specifikationer
- Ø×L:11×35 mm
- Til:Semi-solid substances
- Beskrivelse:Spare pH probe for testo 206 pH2, with NTC, incl. gel storage cap
- Seq:03
- Elektrolyt:Gel
- Temperatur:0...60 °C
- pH:0 - 14