pH probes
Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap, 0 - 14, 0...60 °C, Gel, 15×3,5 mm, Liquids
Katalog # TEST0650.2061
Leverandør: Testo
Gå til:
Product Details & Documents
Specifikationer
- Ø×L:15×3,5 mm
- Til:Liquids
- Beskrivelse:Spare pH probe for testo 206 pH1, with NTC, incl. gel storage cap
- Seq:02
- Elektrolyt:Gel
- Temperatur:0...60 °C
- pH:0 - 14