Single stage gas regulators
Singlel stage gas regulator, for Air
Katalog # REST20667
Leverandør: Restek
Gå til:
Product Details & Documents
These stainless steel regulators are the standard for ultra high purity and corrosion resistant pressure regulation. They are more easily purged of atmospheric components compared to brass gas regulators.
- Use when there is secondary pressure regulation downstream
- Identical gas purity protection as with dual-stage gas regulators
- Especially useful in atmospheres of dry corrosive gases such as hydrogen
- Outlet assembly: 316 L stainless steel diaphragm valve and ¹/₄ tube fitting
Specifikationer
- Beskrivelse:Singlel stage gas regulator, for Air
- Seq:01