Orders LinkContactUsLinkComponent
Single stage gas regulators
Singlel stage gas regulator, for H2, P5
  REST20666
 :   Restek
Single stage gas regulators
  REST20666
 :  Restek
Single stage gas regulators
Single stage gas regulators
Singlel stage gas regulator, for H2, P5
12.500,00 kr
1 * 1 Items
These stainless steel regulators are the standard for ultra high purity and corrosion resistant pressure regulation. They are more easily purged of atmospheric components compared to brass gas regulators.
  • Use when there is secondary pressure regulation downstream
  • Identical gas purity protection as with dual-stage gas regulators
  • Especially useful in atmospheres of dry corrosive gases such as hydrogen
  • Outlet assembly: 316 L stainless steel diaphragm valve and ¹/₄ tube fitting
  • Beskrivelse:
    Singlel stage gas regulator, for H2, P5
  • Seq:
    02