Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment, 1,8 m
Katalog # REST20657
Leverandør: Restek
Gå til:
Product Details & Documents
These are replacement cables for Agilent GCs, integrators and autosamplers.
: Instructions and writing diagrams included.
Specifikationer
- Beskrivelse:Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
- Længde:1,8 m
- Seq:02