Orders LinkContactUsLinkComponent
Capillary nuts
Capillary nuts
Enhanced capillary nut
Catalog # REST20375
Supplier:   Restek
Capillary nuts
Catalog # REST20375
Supplier:  Restek
Capillary nuts
Capillary nuts
Enhanced capillary nut
Price
612,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.
Specifications
  • Beskrivelse:
    Enhanced capillary nut
  • Seq:
    02