Solid phase extraction cartridges, Resprep®, method specific
RDX method, for extraction of explosive compounds from water samples, PP, 6 ml, 500 mg
Katalog # REST26093
Leverandør: Restek
Gå til:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifikationer
- Kapacitet:6 ml
- Beskrivelse:RDX method, for extraction of explosive compounds from water samples
- Seq:04
- Lejevægt:500 mg
- Kolonnemateriale:PP