Solid phase extraction cartridges, Resprep®, method specific
EPA method 548.1, for extraction of endothall from aqueous samples, PP, 6 ml, -
Katalog # REST26063
Leverandør: Restek
Gå til:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifikationer
- Kapacitet:6 ml
- Beskrivelse:EPA method 548.1, for extraction of endothall from aqueous samples
- Seq:03
- Lejevægt:-
- Kolonnemateriale:PP