Solid phase extraction cartridges, Resprep®, method specific
EPA methods 521 and 522, for use in EPA method 521 for nitrosamines in drinking water and EPA method 522 for 1,4-dioxane in drinking water, with activated charcoal packing, PP, 6 ml, 2 g
Katalog # REST26032
Leverandør: Restek
Gå til:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifikationer
- Kapacitet:6 ml
- Beskrivelse:EPA methods 521 and 522, for use in EPA method 521 for nitrosamines in drinking water and EPA method 522 for 1,4-dioxane in drinking water, with activated charcoal packing
- Seq:02
- Lejevægt:2 g
- Kolonnemateriale:PP