Accessories for solid phase extraction cartridges, Resprep®
Frits, PE, 20 µm, for 25 ml tubes (20 mm)
Katalog # REST26020
Leverandør: Restek
Accessories for solid phase extraction cartridges, Resprep®
Katalog # REST26020
Leverandør: Restek
Gå til:
Product Details & Documents
Resprep tubes, frits, caps, and connectors for method development needs.
Specifikationer
- Beskrivelse:Frits, PE, 20 µm, for 25 ml tubes (20 mm)
- Seq:11