Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Fittings, secure-fit

Fittings, secure-fit

Secure-fit fittings, 0,178 mm I-Ø, PEEK, 300 mm
Katalog # REST25237
Leverandør:   Restek
Fittings, secure-fit
Katalog # REST25237
Leverandør:  Restek
Fittings, secure-fit
Fittings, secure-fit

Secure-fit fittings, 0,178 mm I-Ø, PEEK, 300 mm

Price
657,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Secure-Fit provides a good connection between HPLC components for trouble-free chromatography.

  • Consistent, leak-free seal
  • Eliminates the excess dead volume

Internal spring mechanism holds the capillary tubing at the proper depth in the female fitting. This seal is maintained while the nut is finger tightened.

Specifikationer
  • Beskrivelse:
    Secure-fit fittings, 0,178 mm I-Ø
  • Længde:
    300 mm
  • Materiale:
    PEEK