Check valves, ASI
Inlet check valve, sapphire and ruby ball and seat, includes cartridge and housing
Catalog # REST25164
Supplier: Restek
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Inlet check valve, sapphire and ruby ball and seat, includes cartridge and housing
- Seq:10