Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Lab organisers

Book holder, I.Ø 38 mm (large), 114×152×44,5 mm
Katalog # REST25152
Leverandør:   Restek
Lab organisers
Katalog # REST25152
Leverandør:  Restek
Lab organisers
Lab organisers

Book holder, I.Ø 38 mm (large), 114×152×44,5 mm

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These organisers are designed to hold lab supplies.

Please note: Accessories and supplies in photograph are not included.

Specifikationer
  • Beskrivelse:
    Book holder, I.Ø 38 mm (large), 114×152×44,5 mm
  • Seq:
    06
  • Cat. No.:
    REST25152
  • Dispenser type:
    Bænk top