Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lab organisers

Lab organisers

Book holder, I.Ø 19 mm (small), 114×152×31,8 mm
Catalog # REST25151
Supplier:   Restek
Lab organisers
Catalog # REST25151
Supplier:  Restek
Lab organisers
Lab organisers

Book holder, I.Ø 19 mm (small), 114×152×31,8 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

These organisers are designed to hold lab supplies.

Please note: Accessories and supplies in photograph are not included.

Specifications
  • Beskrivelse:
    Book holder, I.Ø 19 mm (small), 114×152×31,8 mm
  • Seq:
    05
  • Cat. No.:
    REST25151
  • Dispenser type:
    Bænk top
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.