Precision sampling valves for vials, Mininert®
Septum insertion tool
Katalog # REST24907
Leverandør: Restek
Gå til:
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
- Available for screw cap and crimp top vials
- Threaded flange used to secure tight fit
Specifikationer
- Beskrivelse:Septum insertion tool
- Seq:08
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Replacement septa
Produktinformation
- Katalog #TilgængelighedPrice
- Septum insertion tool