Orders LinkContactUsLinkComponent
Precision sampling valves for vials, Mininert®
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 13 mm/425 thread
Katalog # REST24900
Leverandør:   Restek
Precision sampling valves for vials, Mininert®
Katalog # REST24900
Leverandør:  Restek
Precision sampling valves for vials, Mininert®
Precision sampling valves for vials, Mininert®
Mininert® precision sampling valves, for screw-thread vials, 13 mm/425 thread
Price
1.610,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
Mininert® valves are very convenient for repetitive sampling and limit content exposure to the silicon septum. The crimp-top valve for 20 mm ID glassware slides into the neck of the vial.
  • Available for screw cap and crimp top vials
  • Threaded flange used to secure tight fit
Specifikationer
  • Beskrivelse:
    Mininert® precision sampling valves, for screw-thread vials, 13 mm/425 thread
  • Seq:
    01
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Replacement septa
    Replacement septa
    REST24906
    Log ind for tilgængelighed
    1 * 50 Items - 485,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Septum insertion tool
    Septum insertion tool
    REST24907
    Log ind for tilgængelighed
    1 * 1 Items - 441,00 kr