Crimp top vials with inserts
Vials with 100 µl glass insert, 2,00 ml, Plast, klar, No
Katalog # REST24653
Leverandør: Restek
Gå til:
Product Details & Documents
Crimp-top plastic vial with 100 µl glass or PP fused insert.
Specifikationer
- Kasket inkluderet:No
- Kapacitet:2,00 ml
- Farve:klar
- Beskrivelse:Vials with 100 µl glass insert
- Materiale:Plast