Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Vacuum gauges

Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 6,35 mm NTP fitting
Katalog # REST24270
Leverandør:   Restek
Vacuum gauges
Katalog # REST24270
Leverandør:  Restek
Vacuum gauges
Vacuum gauges

Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 6,35 mm NTP fitting

Price
1.420,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These vacuum gauges are made of stainless steel wetted surfaces with rear mount.

Suitable for passive air sampling kits.

Specifikationer
  • Beskrivelse:
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 6,35 mm NTP fitting
  • Seq:
    02