Flow controllers, Veriflo
Rustfrit stål, Flow controller, without orifice
Katalog # REST24239
Leverandør: Restek
Gå til:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Specifikationer
- Beskrivelse:Flow controller, without orifice
- Materiale:Rustfrit stål
- Seq:01