Flow controllers, Veriflo
Siltek® behandelter Edelstahl, Flow controller, without orifice
Katalog # REST24238
Leverandør: Restek
Gå til:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Specifikationer
- Beskrivelse:Flow controller, without orifice
- Materiale:Siltek® behandelter Edelstahl
- Seq:02