Flow controllers, Veriflo
0,5 – 2,0 ml/min, Rustfrit stål, Flow controller, 0,02 mm
Katalog # REST24229
Leverandør: Restek
Gå til:
Product Details & Documents
Veriflo controllers can be used with passive air samplers in maintaining a constant flow rate.
Entire sample flow path is made of stainless steel or siltek® treated for maximum inertness.
Specifikationer
- Ø:0,02 mm
- Maksimal flowhastighed:0,5 – 2,0 ml/min
- Beskrivelse:Flow controller
- Materiale:Rustfrit stål
- Seq:03