Orders LinkContactUsLinkComponent
Miniature air sampling canisters
Miniature canister, 400 ml
Catalog # REST24189
Supplier:   Restek
Miniature air sampling canisters
Catalog # REST24189
Supplier:  Restek
Miniature air sampling canisters
Miniature air sampling canisters
Miniature canister, 400 ml
Price
4.200,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These small canisters are designed for controlled sampling, as an alternative to tube and pump samplers.
  • Treated/electropolished stainless steel
  • Equipped with quick-connect fitting for sampling and analysis instruments
  • Canister with about 1000 cc suitable for sampling volatile organic compounds in air
Suitable for indoor air, personal, emergency response, or soil gas sampling.
 : NOTE: Suitable for low pressure application. Should not exceed 40 psig (2,75 bar).
Specifications
  • Beskrivelse:
    Miniature canister, 400 ml
  • Seq:
    201