Passive air sampling kits
Replacement frit filters, 2 µm, —, Siltek® behandelter Edelstahl
Katalog # REST24171
Leverandør: Restek
Gå til:
Product Details & Documents
This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
- Different orifice sizes for sampling from four minutes to ten hours
- Compression sample inlet allows easy connection
- Canister valve connected to a sample port tubing
- Stainless steel tee with orifice and vaccum gauge
- Frit filter for insertion into a compression sample inlet
Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
Bestillingsoplysninger: Canisters and replacement tees and orifices can be ordered separately.
Specifikationer
- Beskrivelse:Replacement frit filters, 2 µm
- Materiale:Siltek® behandelter Edelstahl
- Seq:902
- Luftflow:—