Solid phase extraction cartridges, Resprep®, method specific
Organo tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bed, PP, 60 ml, -
Katalog # REST24049
Leverandør: Restek
Gå til:
Product Details & Documents
Cartridges have been specifically designed to provide consistent and reproducible results for the method or application stated.
Specifikationer
- Kapacitet:60 ml
- Beskrivelse:Organo tin method, for high capacity clean-up of butyl and phenyl tin compounds from soil, water and biota. Mixed bed
- Seq:06
- Lejevægt:-
- Kolonnemateriale:PP