Solid phase extraction cartridges, Resprep®
SPE Florisil cartridges, with PTFE frits, Patron, 3 ml, 500 mg
Katalog # REST24032
Leverandør: Restek
Gå til:
Product Details & Documents
These cartridges are manufactured using high density polypropylene and have polyethylene frits unless otherwise noted. Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices, such as organic solvents (e.g., polar contaminants from sample extracts).
Cartridges may be processed by any one or all of these techniques: positive pressure, sidearm flask, centrifuge, or vacuum manifold.
Specifikationer
- Kolonne form:Patron
- Kapacitet:3 ml
- Beskrivelse:SPE Florisil cartridges, with PTFE frits
- Seq:02
- Lejevægt:500 mg