Solid phase extraction disks, Resprep®
Solid phase extraction disks, Resprep® C18, 47 mm
Catalog # REST24004
Supplier: Restek
Jump to:
Product Details & Documents
These glass fiber disks are embedded with C18 or C8 bonded silica. Deep-pore design reduces clogging and allows faster flow rates. Semivolatile organic compounds can be etracted.
Certifications: Meet the requirements for EPA methods 525.1, 506, 550.1 and 549.1.
Specifications
- Ø:47 mm
- Beskrivelse:Solid phase extraction disks, Resprep® C18
- Seq:02