Orders LinkContactUsLinkComponent
Rethreading tool
Rethreading tool
Rethreading tool, for 1/4" compression fitting (Agilent split/splitless injection ports)
Katalog # REST23018
Leverandør:   Restek
Rethreading tool
Katalog # REST23018
Leverandør:  Restek
Rethreading tool
Rethreading tool
Rethreading tool, for 1/4" compression fitting (Agilent split/splitless injection ports)
Price
836,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These rethreading tools are suitable for repairing worn or damaged threads that come as a result of consistent use and temperature changes.
  • Multiple uses (injection ports, fittings, etc.)
  • Built-in guide prevents cross-threading
Repeated installation, removal and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Specifikationer
  • Beskrivelse:
    Rethreading tool, for 1/4" compression fitting (Agilent split/splitless injection ports)
  • Seq:
    03