Orders LinkContactUsLinkComponent
Capillary nuts
Capillary nuts
Capillary nut
Katalog # REST22688
Leverandør:   Restek
Capillary nuts
Katalog # REST22688
Leverandør:  Restek
Capillary nuts
Capillary nuts
Capillary nut
Price
620,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
These capillary nuts are ideal for Shimadzu 17A, 2010, and 2014 GCs.
The enhanced capillary nut design eliminates the slot, increasing lifetime and durability.
Specifikationer
  • Beskrivelse:
    Capillary nut
  • Seq:
    01