Injector adapters
Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts, PE-style capillary nuts
Katalog # REST22320
Leverandør: Restek
Gå til:
Product Details & Documents
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Specifikationer
- Til:PE-style capillary nuts
- Beskrivelse:Injector adapter for PerkinElmer CAP injector, stainless steel, for PE-style capillary nuts
- Seq:02