Injector adapters
Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts, 1/16" compression-style nuts
Katalog # REST22319
Leverandør: Restek
Gå til:
Product Details & Documents
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Specifikationer
- Til:1/16" compression-style nuts
- Beskrivelse:Injector adapter for PerkinElmer CAP injector, Siltek®-treated, for 1/16" compression-style nuts
- Seq:03