Orders LinkContactUsLinkComponent
Protocol wall mounts for gas regulators
Protocol wall mounts for gas regulators
Protocol station, chrome-plated brass, CGA 590 (Air) fitting
Katalog # REST21349
Leverandør:   Restek
Protocol wall mounts for gas regulators
Katalog # REST21349
Leverandør:  Restek
Protocol wall mounts for gas regulators
Protocol wall mounts for gas regulators
Protocol station, chrome-plated brass, CGA 590 (Air) fitting
Price
6.530,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Ease of use
  • Prevents gas regulator damage and improves safety
Either chrome-plated brass or 316 stainless steel option is complete with a 0,9m, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Specifikationer
  • Beskrivelse:
    Protocol station, chrome-plated brass, CGA 590 (Air) fitting
  • Seq:
    03