Orders LinkContactUsLinkComponent
Jumbo syringes
Jumbo syringes
2000 ml, 2000MAR-LL-GT
Katalog # REST21277
Leverandør:   Restek
Jumbo syringes
Katalog # REST21277
Leverandør:  Restek
Jumbo syringes
Jumbo syringes
2000 ml, 2000MAR-LL-GT
Price
12.720,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
  • Central port with a Luer lock fitting
  • Secondary port with septum nut
  • Detachable plunger stem
  • Easy sample storage
An adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time.
Specifikationer
  • Model:
    2000MAR-LL-GT
  • Volumen:
    2000 ml
  • Seq:
    03
Accessories
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Syringe O-ring
    Syringe O-ring
    REST21278
    500 ml syringes
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Til
    Tilgængelighed
    Price
  • Syringe O-ring
    Syringe O-ring
    REST21279
    1000 ml syringes