Caps for screw thread vials
Open-hole caps, PP, black
Katalog # REST21176
Leverandør: Restek
Gå til:
Product Details & Documents
Black PP, open hole caps fit all 2,0 ml, 12×32 mm, screw-thread 8 mm/425 vial-based autosamplers.
Specifikationer
- Beskrivelse:Open-hole caps, PP, black
- Seq:01