GC columns, Rt®-608 (packed)
1,5% Rt-608 with PE sigma column configuration, Kapillær, 2,00 mm, 2 m
REST80221-830
Optimised for pesticide analysis by US EPA Method 608.
- Kolonne form:Kapillær
- Ø indv.:2,00 mm
- Beskrivelse:1,5% Rt-608 with PE sigma column configuration
- Længde:2 m