Orders LinkContactUsLinkComponent
GC columns, Rt®-608 (packed)
GC columns, Rt®-608 (packed)
1,5% Rt-608 with agilent column configuration, Kapillær, 2,00 mm, 2 m
Katalog # REST80221-810
Leverandør:   Restek
GC columns, Rt®-608 (packed)
Katalog # REST80221-810
Leverandør:  Restek
GC columns, Rt®-608 (packed)
GC columns, Rt®-608 (packed)
1,5% Rt-608 with agilent column configuration, Kapillær, 2,00 mm, 2 m
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Optimised for pesticide analysis by US EPA Method 608.
Specifikationer
  • Kolonne form:
    Kapillær
  • Ø indv.:
    2,00 mm
  • Beskrivelse:
    1,5% Rt-608 with agilent column configuration
  • Længde:
    2 m