GC columns, Rt®-608 (packed)
1,5% Rt-608 with ON COL/ Tracor 550 column configuration, Kapillær, 4,00 mm, 2 m
Catalog # REST80220-870
Jump to:
Discontinued
Product Details & Documents
Optimised for pesticide analysis by US EPA Method 608.
Specifications
- Kolonne form:Kapillær
- Ø indv.:4,00 mm
- Beskrivelse:1,5% Rt-608 with ON COL/ Tracor 550 column configuration
- Længde:2 m