GC columns, Rt®-608 (packed)
1,5% Rt-608 with 4,76 mm general column configuration, Kapillær, 4,00 mm, 2 m
Katalog # REST80220-800
Gå til:
Discontinued
Product Details & Documents
Optimised for pesticide analysis by US EPA Method 608.
Specifikationer
- Kolonne form:Kapillær
- Ø indv.:4,00 mm
- Beskrivelse:1,5% Rt-608 with 4,76 mm general column configuration
- Længde:2 m