Orders LinkContactUsLinkComponent
Columns, MXT®-5
Columns, MXT®-5
3,5" coil, Kapillær, MXT®-5, 1,00 µm, 0,53 mm, 30 m
Catalog # REST70255-273
Supplier:   Restek
Columns, MXT®-5
Catalog # REST70255-273
Supplier:  Restek
Columns, MXT®-5
Columns, MXT®-5
3,5" coil, Kapillær, MXT®-5, 1,00 µm, 0,53 mm, 30 m
Price
11.050,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These MXT-5 columns are Siltek-treated stainless steel with low-polarity phase, crossbond diphenyl dimethyl polysiloxane stationary phase. All residual catalysts and low molecular weight fragments are removed from the MXT-5 polymer, providing a tight monomodal distribution and extremely low bleed.
  • Equivalent to USP G27, G36 phases
  • 4,5 inch standard coil diameter
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners, essential oils, and semivolatiles.
Specifications
  • Kolonne form:
    Kapillær
  • Filmtykkelse:
    1,00 µm
  • Ø indv.:
    0,53 mm
  • Beskrivelse:
    3,5" coil
  • Længde:
    30 m
  • Seq:
    30
  • Fase:
    MXT®-5