Lids for 96-well microplates
96-well microplate lid, high profile (9 mm), with condensation rings, klar, Ikke steril
736-6552
These high profile lids, height 9 mm, are designed to protect against contamination and evaporation during sample storage and cell culture. Lids without condensation rings are mainly used with non tissue culture treated microplates. Lids with condensation rings are recommended for cell culture applications, since they inhibit cross contamination.
- Sterilitet:Ikke steril
- Farve:klar
- Beskrivelse:96-well microplate lid, high profile (9 mm), with condensation rings