Wound cleansers
Dispenser til Savett-renseservietter, fyldt
Katalog # 118-0025
Leverandør: Orkla Care AB / Cederroth
Gå til:
Product Details & Documents
Wound cleansers do away with cotton wool and bottles of antiseptic. These sterile, non woven tissues, which are saturated in a wound cleansing fluid (0,9% NaCl), are intended for the mechanical cleansing of minor wounds when clean tap water is not available.
- Tissues are packed in sealed sachets, which prevent leakage and guarantee sterility
- Savett sachets are packed in a refill box
- Salvequick cleansers fit into first-aid-stations
Documents
VWR® for Lab Safety
Produktlitteratur
Specifikationer
- Beskrivelse:Dispenser til Savett-renseservietter, fyldt