Tanks for water baths
Plexiglas® tank with inclined bottom, 430×130×150 mm
Katalog # HECH43187003
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Tubs made of Plexiglas®, available with or without sloping base. Sloping base facilitates circulation of the liquid.
For use with water bath thermostats.
Specifikationer
- Beskrivelse:Plexiglas® tank with inclined bottom, 430×130×150 mm
- Seq:04