Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® ECO pH 6,1 - 8,4, refill pack, test no.5-70; Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation, Reagents for 150 tests in a box, pH: 6.1 - 8.4, Mixed indicator, PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance, 170×115×70 mm (including packaging), Hazardous material: No, 150, 1 KIT
Katalog # MANA931270
Leverandør:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, pH
Katalog # MANA931270
Leverandør:  MACHEREY-NAGEL
CAS nummer:  
VISOCOLOR® Colorimetric Test, pH
VISOCOLOR® Colorimetric Test, pH

VISOCOLOR® ECO pH 6,1 - 8,4, refill pack, test no.5-70; Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation, Reagents for 150 tests in a box, pH: 6.1 - 8.4, Mixed indicator, PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance, 170×115×70 mm (including packaging), Hazardous material: No, 150, 1 KIT

CAS nummer:  
Price
688,00 kr
Log ind for tilgængelighed
1 * 1 Each
Specifikationer

Specifikationer

  • Kemisk metode:
    Mixed indicator
  • Pakn:
    1 KIT
  • Beskrivelse:
    VISOCOLOR® ECO pH 6,1 - 8,4, refill pack, test no.5-70; Measuring range on PF-12Plus. Range on other photometers can be different. Only for photometric evaluation
  • Dimensioner:
    170×115×70 mm (including packaging)
  • Model:
    Hazardous material: No
  • Til enhed:
    PF-12Plus; PF-3 Fish; PF-3 Pool; PF-3 Drinking Water; Advance
  • Test pr. sæt:
    150
  • Område:
    pH: 6.1 - 8.4
  • Indhold:
    Reagents for 150 tests in a box
  • Cat. No.:
    MANA931270
  • Platform:
    Photometric test kits
  • Holdbarhed (måneder):
    18
  • Vægt:
    105,91 g (including packaging)
  • Model:
    MANA931270
  • Opbevaringstemperatur:
    15…25 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.