Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
33× 80 mm, 2,0 mm
Katalog # MANA645951
Leverandør:   MACHEREY-NAGEL
Extraction thimbles
Katalog # MANA645951
Leverandør:  MACHEREY-NAGEL
Extraction thimbles
Extraction thimbles
33× 80 mm, 2,0 mm
Price
1.040,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
  • Fat determination in food and feed
  • Determination of binder content in asphalt
  • Analysis of soil contaminants
  • Determination of additives in rubber and elastomers
Specifikationer
  • Indv. Ø×H:
    33× 80 mm
  • Seq:
    24
  • Tykkelse:
    2,0 mm